This item is now discontinued. Please browse similar items
Perfex 9524 Heavy-Duty Floor Squeegee, twin-mounted PVC blades, 24"W
Representative image only.
Perfex Mfr # 9524 – Item # RK-86517-00
PVC blades are non-conductive and will not absorb bacteria, liquids, or odors
SPECS & DESCRIPTION
Specifications & Description
  • ColorWhite
  • Width (in)24
  • DescriptionHeavy-Duty Floor Squeegee, twin-mounted PVC blades, 24\"W
More About this Item
100% polymer construction is chemically-resistant, non-sparking, and will not rust or corrode. Self-locking connection system quickly threads handle into connector for a tight fit. Select from 24" or 30" blades to adjust for space. Lightweight, durable squeegees are unaffected by water, grease, sanitizers, solvents, and petroleum products. Rinses clean and dries quickly.
Approval:Material approved by USDA. FDA compliant.