This item is now discontinued. Please browse similar items
PCRmax Sealing Foil for 96- and 384-Well PCR/qPCR Plates, transparent, self-adhesive, PET; 100/pk
Representative image only.
PCRmax Item # RK-67104-51
Reduce evaporation losses
  • Free of RNase, DNase and endotoxins
  • Thermostable and pressure resistant—ideal for use with thermal cyclers with heated lids
  • Easily strips off without leaving a sticky residue on the plate
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPET
  • DescriptionSealing Foil for 96- and 384-Well PCR/qPCR Plates, transparent, self-adhesive, PET; 100/pk
Key Features
  • Free of RNase, DNase and endotoxins
  • Thermostable and pressure resistant—ideal for use with thermal cyclers with heated lids
  • Easily strips off without leaving a sticky residue on the plate