Parker Hannifin 2 BZ-SS-C Compression Fitting Ferrule Nut, 316SS, 1/8" OD; 1/EA
Representative image only.
Parker Hannifin Mfr # 2 BZ-SS-C – Item # RK-03302-46
$10.17CAD / Each
BUY MORE & SAVE
  • 1-9$10.17CAD / Each
  • 10-49$9.15CAD / Each
  • 50-99$8.64CAD / Each
  • 100+$8.14CAD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialStainless Steel
  • Connection TypeQuick-Disconnect
  • DescriptionCompression Fitting Ferrule Nut, 316SS, 1/8" OD; 1/EA
More About this Item
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
Compression nuts are required to create a proper seal.