This item is now discontinued. Please browse similar items
Pacific Scientific-OECO 4190 Handheld Gaussmeter, Tr-Axis,<1% Accrcy Includes F20 Option
Representative image only.
Pacific Scientific-OECO Mfr # 4190 – Item # RK-20001-34
The 4100 Series Handheld Gaussmeter accurately measures extremely low frequency magnetic fields generated by electrical equipment; ideal for home use
  • Low cost, small, lightweight, and portable
  • High accuracy (< 1%, Model 4190) ± 1% +1 digit
  • 0.1 mG resolution
  • Gauss or Tesla display options
  • Min/Max hold and relative measurement capabilities (4190 only)
  • Selectable X, Y, or Z Axis plus vector summation
  • Waveform and RMS analog output (4190 only)
  • Universal Serial Bus Interface, Remote Operation and USB remote on both 4180 and 4190
  • Unit can be powered from 4 AAA batteries, external DC supply, or USB Bus Power (4190 only)
  • Low battery detection
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionHandheld Gaussmeter, Tr-Axis,<1% Accrcy Includes F20 Option
  • Warranty1 Year
1 Year Warranty
Key Features
  • Low cost, small, lightweight, and portable
  • High accuracy (< 1%, Model 4190) ± 1% +1 digit
  • 0.1 mG resolution
  • Gauss or Tesla display options
  • Min/Max hold and relative measurement capabilities (4190 only)
  • Selectable X, Y, or Z Axis plus vector summation
  • Waveform and RMS analog output (4190 only)
  • Universal Serial Bus Interface, Remote Operation and USB remote on both 4180 and 4190
  • Unit can be powered from 4 AAA batteries, external DC supply, or USB Bus Power (4190 only)
  • Low battery detection