This item is now discontinued. Please browse similar items
Opto 22 PACPROJECTBAS PAC Project Basic Software Suite
Representative image only.
Opto 22 Mfr # PACPROJECTBAS – Item # RK-20011-77
  • FEATURES - R Series
  • Convenient PAC package for small or distributed applications
  • Compact rack-mounted industrial controller with I/O processor included (PAC-R-series)
  • Two independent 10/100 Mbps Ethernet network interfaces
  • One RS-232 port with handshaking suitable for modern communications using PPP
  • Full-featured control and HMI software included with purchase.
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionPAC Project Basic Software Suite
Key Features
  • FEATURES - R Series
  • Convenient PAC package for small or distributed applications
  • Compact rack-mounted industrial controller with I/O processor included (PAC-R-series)
  • Two independent 10/100 Mbps Ethernet network interfaces
  • One RS-232 port with handshaking suitable for modern communications using PPP
  • Full-featured control and HMI software included with purchase.
More About this Item
The SNAP PAC System is an integrated system of hardware and software for industrial control remote monitoring and data acquisition applications
  • Standalone controller suitable for larger and more complex applications
  • Up to 32 ioControl flowcharts running simultaneously
  • Two 10/100 Mbps Ethernet interfaces for host and I/O communication over standard Ethernet networks
  • One RS-485 serial port for connection to Opto 22 Mistic I/O units
  • Two RS-232 serial ports (one with full handshaking) for PPP host
    connection or direct connections to serial devices
  • Multiple protocol support:TCP/IP SNMP SMTP FTP OptoMMP™
  • FTP server/client capability; includes file system