This item is now discontinued. Please browse similar items
Omegadyne LCKD1000 Subminiature Compression Load Cells, 1000 LB capacity
Representative image only.
Newport Mfr # LCKD1000 – Item # RK-59890-97
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (lbs)1000
  • Capacity (kg)455
  • DescriptionSubminiature Compression Load Cells, 1000 LB capacity
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Key Features
  • 1 year warranty
More About this Item
Sub-miniature, very low profileThese sub-miniature compression load cells are designed to measure load ranges from 1 kg to 1000 lb. Rugged all stainless steeel construction and high-performance strain gauges assure superior linearity and stability. These cells are designed to be mounted on a smooth, flat surface.