This item is now discontinued. Please browse similar items
Omegadyne LCHD-10 Load Cell10LB.Cap Low Profile Pancake Tension And Compression Load Cell
Representative image only.
Omegadyne Mfr # LCHD-10 – Item # RK-93955-76
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (lbs)10
  • Capacity (kg)4.5
  • Output=25 lb: 2 mV/V ±0.25%; =50 lb: 3 mV/V ±0.25%
  • DescriptionLoad Cell10LB.Cap Low Profile Pancake Tension And Compression Load Cell
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Low profile pancake style
  • 0.25% interchangeability for scale applications
  • All stainless steel construction
  • Heavy-Duty design
  • High accuracy
  • Low profile for easy installation