This item is now discontinued. Please browse similar items
Omegadyne LC703-10 Load Cell10LB. Capacity Mini Low Pr Tension Link Load Cell
Representative image only.
Omegadyne Mfr # LC703-10 – Item # RK-93955-42
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (lbs)10
  • Capacity (kg)4.5
  • Output2 mV/V nominal
  • DescriptionLoad Cell10LB. Capacity Mini Low Pr Tension Link Load Cell
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Miniature Low Profile Tension Links These tension / compression load cells have an extremely low profile. Ranges above 100 lb are stainless steel; ranges below 100 lb are aluminum. The low profile and rugged design make these LC703’s suitable for many industrial applications, including robotics, automated weighing systems, and batch-process control systems.