This item is now discontinued. Please browse similar items
Oakton SaltSingles NaCl Calibration Standard Solution; 20/Pk
Representative image only.
Oakton Item # RK-35653-15
Calibration standard solution for salt meters
  • Single-use 20-mL pouches are sealed to ensure freshness—take these convenient solutions anywhere!
  • Slim desgin slips into pouches and cases
  • Calibration solution for salt; 3000 ppm (3.00 ppt)
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Volume (mL)20
  • Volume (oz)0.7
  • Volume20 pouches
  • Conductivity-to-TDS Calibration Values (ppm NaCl)3000
  • Description3000 ppm salt singles, NaCl calibration packets
  • Warranty1 year
1 Year Warranty
Key Features
  • Single-use 20-mL pouches are sealed to ensure freshness—take these convenient solutions anywhere!
  • Slim desgin slips into pouches and cases
  • Calibration solution for salt; 3000 ppm (3.00 ppt)
  • 1 year warranty
More About this Item
No need for separate calibration containers—tear open the pouch, insert the probe or salt tester, and calibrate your meter. One-time-use solution pack makes calibrating in the field simple.
Calibration pouches include a NIST-traceable calibration report.
Certificate of Analysis
Enter lot number
or click to view a Sample C of A