Oakton Replacement Electrode Arm for Meters
Representative image only.
Oakton Item # RK-35420-54
Replace or add an electrode holder arm to your Oakton benchtop meters
  • Electrode holder arm can be mounted on either side of meter
  • Ability to add two electrode arms; one on each side of the meter
  • 1 year warranty
$321.94CAD / Each
Include Benchtop Meter!
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionReplacement Electrode Arm for Meters
  • Warranty1 year
1 Year Warranty
Key Features
  • Electrode holder arm can be mounted on either side of meter
  • Ability to add two electrode arms; one on each side of the meter
  • 1 year warranty
More About this Item
Electrode holder with swivel arm allows convenient hands-free operation. Hold up to four standard size probes and ATC probe. Arm allows you to move electrode up and down and side to side while keeping electrode at a constant verticle angle.