Oakton pHTestr® 5 Waterproof Pocket Tester with pH Buffer Pouches

Representative image only.
Oakton Item # RK-35634-66
Save money by replacing sensor, not the tester
  • Large easy-to-read LCD features three-color backlight
  • Simple to calibrate with automatic standard recognition
  • Rugged IP67-rated housing is waterproof, dustproof, and floats in water with sensor cap on
  • Leakproof cap with fill line functions as a sample/calibration and hydrating cup
  • Secure—attach a lanyard to prevent losing or dropping in the field
  • Auto shutoff extends battery life
  • Bundled kit for convenience and savings!
  • Economical pH buffer pouches are accurate and convenient
  • 2 year warranty

PRODUCT OPTIONS

$294.68CAD / Pkg of 1
Include Accessories Today!

SPECS & DESCRIPTION

Specifications & Description

  • Display typeDual-line LCD
  • Min pH (pH)-2
  • Max pH (pH)16
  • pH Resolution0.01 pH
  • pH Accuracy±0.01 pH ±1 digit
  • Buffer RecognitionNIST and USA
  • Calibration1 to 3 points
  • Min Temperature (° C)0
  • Max Temperature (° C)50
  • Min Temperature (° F)32
  • Max Temperature (° F)122
  • Temperature Resolution0.1°F/°C
  • Temperature Accuracy±0.9°F (0.5°C)
  • Temperature compensationAutomatic
  • WaterproofYes
  • Battery4 AAA (included)
  • Battery life (hr)400
  • Length (cm)17.8
  • Width (cm)4
  • Height (cm)4
  • Width (in)1 1/2
  • Length (in)7
  • Height (in)1 1/2
  • DescriptionWaterproof Pocket Tester with pH Buffer Pouches
Two year warrantyConformite EuropeenneIP 67

Includes

Pack of 20 assorted (4, 7, and 10) buffer pouches, plastic storage case, lanyard, and four AAA batteries.

Key Features

  • Large easy-to-read LCD features three-color backlight
  • Simple to calibrate with automatic standard recognition
  • Rugged IP67-rated housing is waterproof, dustproof, and floats in water with sensor cap on
  • Leakproof cap with fill line functions as a sample/calibration and hydrating cup
  • Secure—attach a lanyard to prevent losing or dropping in the field
  • Auto shutoff extends battery life
  • Bundled kit for convenience and savings!
  • Economical pH buffer pouches are accurate and convenient
  • 2 year warranty

More About this Item

These pocket-sized testers have a replaceable single-junction sensor which can be easily switched out, allowing you to save money by using the tester body over and over again. A large LCD makes viewing measurements easy and features a three-color backlight indicating operational mode—blue for measurement, green for calibration and red for alarm condition.
Additional features include hold function to lock in measurements, self-diagnostic error message, and status icons for measurement stability and low battery voltage. The IP67-rated housing features an easy-to-grip design and floats if accidentally dropped in water when sensor cap is on. Powered by four AAA batteries, the testers can operate up to 400 hours continuously. To conserve power, the unit will auto power-off after 8 minutes of nonuse.

Buffers are traceable and offer ±0.01 accuracy at 77°F (25°C).

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat