Oakton pH Indicator Strips, 0 to 14, 100/pk
Representative image only.
Oakton Item # RK-35850-02
Vivid and easy-to-distinguish colors at every pH interval
  • 3 month warranty
PRODUCT OPTIONS
$44.94CAD / Pkg of 100
Include Beakers Today!
SPECS & DESCRIPTION
Specifications & Description
  • TestpH
  • Measurement Points0 to 14 pH in 1 pH unit increments
  • Reagent PlatformStrips
  • Tests Per Kit100
  • DescriptionpH Indicator Strips, 0 to 14, 100/pk
  • Warranty3 month
3 Month Warranty
Key Features
  • 3 month warranty
More About this Item
Convenient flip-top vials include color comparator chart. Read results immediately after exposing test area to the solution. Strips measure 2-3/4"L x 1/4"W (7 x 2 cm). Each vial contains 100 strips.