Oakton pH electrode, epoxy-body, spear tip, double-junction, BNC
Representative image only.
Cole-Parmer Item # RK-59002-24
Unique probes for special applications
  • Combine quality with durability
  • Eposy-body housing resists impact
  • Compatible with most pH meters
  • 1 year warranty
PRODUCT OPTIONS
$259.32CAD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Electrode TypeDouble junction
  • Electrode Body MaterialEpoxy
  • Reference ElectrolyteAg/AgCl
  • Min pH (pH)14
  • Min Temperature (° C)0
  • Max Temperature (° C)80
  • ConnectorBNC
  • Cable Length (m)1
  • DescriptionpH electrode, epoxy-body, spear tip, double-junction, BNC
  • Warranty1 year
1 Year Warranty
Key Features
  • Combine quality with durability
  • Eposy-body housing resists impact
  • Compatible with most pH meters
  • 1 year warranty
More About this Item
These quality electrodes have a no maintenance sealed gel reference. All electrodes have a Ag/AgCl reference cell.
Spear-tip has a double-junction reference to work with contaminants like TRIS, proteins, or sulfides.