Oakton pH Calibration Buffer Pouches, pH 4.01; 20/Pk
Representative image only.
Oakton Item # RK-35653-01
Economical pouches are accurate and convenient
  • Single-use pouch—just tear, dip, calibrate
  • Standardized to NIST-traceable pH references
  • Sold in pack of 20 pouches
  • 1 year warranty
PRODUCT OPTIONS
$85.46CAD / Pkg of 20
Need other buffer values?
Oakton® pH Buffer Pouches
$85.46 - $94.30CAD / Pkg of 20
SPECS & DESCRIPTION
Specifications & Description
  • Buffer Kit Values4.01
  • Volume (mL)20
  • DescriptionpH Calibration Buffer Pouches, pH 4.01, 20/Pk
  • Warranty1 year
1 Year Warranty
Key Features
  • Single-use pouch—just tear, dip, calibrate
  • Standardized to NIST-traceable pH references
  • Sold in pack of 20 pouches
  • 1 year warranty
More About this Item
Buffers are traceable and offer ±0.01 accuracy at 77°F (25°C). Pack of 20 pouches.
Certificate of Analysis
Enter lot number
or click to view a Sample C of A