Oakton PH 550 Benchtop pH Meter Kit

Representative image only.
Oakton Item # RK-35419-30
Economical meter for benchtop pH measurement
  • Automatically recognizes calibration solutions
  • Compact design saves valuable benchtop space
  • Data logging allows you to store and recall up to 50 groups of data
  • IP54-rated housing is dust- and spill-proof
  • 3 year warranty

PRODUCT OPTIONS

$779.42CAD / Each

$242.37 - $349.19CAD / Each

Time to recalibrate? Click on the calibration service to view details, then order.
Include Storage Solution!

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypeBenchtop
  • Display typeLCD
  • Min pH (pH)0
  • Max pH (pH)14
  • pH Resolution0.01/0.1
  • pH Accuracy±0.01 pH ±1 digit
  • Buffer RecognitionNIST and USA
  • CalibrationUP to 3 points
  • Min mV (mV)-1999
  • Max mV (mV)1999
  • mV Resolution1
  • Min Temperature (° C)0
  • Max Temperature (° C)100
  • Min Temperature (° F)32
  • Max Temperature (° F)212
  • Temperature Resolution0.1/1°F (0.1°C)
  • Temperature Accuracy±0.5°C ±1 digit
  • Temperature compensation Automatic or manual from 32 to 122°F (0 to 50°C)
  • Data Logging (points)50 groups of data
  • Length (cm)23.32
  • Width (cm)15.24
  • Height (cm)6.35
  • Width (in)6
  • Length (in)8
  • Height (in)2 1/2
  • DescriptionBenchtop pH Meter Kit with Probe and Stand
  • Warranty3 year
3 Year WarrantyConformite EuropeenneIP54 Rated

Includes

Plastic 3-in-1 combination pH electrode, electrode holder, pH standard buffer solutions (4.00/7.00/10.01 pH, 50 mL each), and 9 VAC power adapter.

Key Features

  • Automatically recognizes calibration solutions
  • Compact design saves valuable benchtop space
  • Data logging allows you to store and recall up to 50 groups of data
  • IP54-rated housing is dust- and spill-proof
  • 3 year warranty

More About this Item

The PH 550 benchtop pH meter features a microprocessor chip which provides automatic temperature compensation, data storage, parameter setup, and Max/Min reading display. The meter offers a pH range from 0 to 14 and a three-point automatic calibration, with the ability to recognize standard buffer solutions (USA or NIST series). The advanced digital processing technology improves meter response time and measuring accuracy. A stable reading display icon appears when the measuring value is stable.

The 3-in-1 combination pH electrode measures pH and temperature simultaneously. The compact meter includes a detachable electrode holder and three bottles of pH buffer solution. It has a dustproof and spillproof rating of IP54. Connectors are protected by silicone seal caps.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat