Oakton PD 450 Waterproof pH/DO Portable Meter with Probes

Representative image only.
Oakton Item # RK-35632-30
Obtain pH and dissolved oxygen readings from one meter
  • Optical DO sensor eliminates costly, tiresome membranes and solutions
  • Durable waterproof and dustproof design with IP67 rating
  • Barometric pressure and salinity correction
  • Memory for up to 500 data sets with USB or RS-232 output to PC
  • Built-in stand allows for easy benchtop use
  • 500 hours battery life, or optional universal power adapter
  • 3 year warranty

PRODUCT OPTIONS

$4,206.53CAD / Each

$245.32 - $365.40CAD / Each

Time to recalibrate? Click on the calibration service to view details, then order.
Include Calibration Solution!

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypePortable
  • Display typeBacklit LCD
  • Min pH (pH)-2
  • Max pH (pH)16
  • pH Resolution0.01
  • pH Accuracy±0.01
  • Buffer RecognitionDIN, NIST, USA, and custom
  • CalibrationUp to 6 points
  • Min mV (mV)-2000
  • Max mV (mV)2000
  • mV Resolution0.01 mV
  • mV Accuracy±0.5% FS
  • Min Dissolved Oxygen0 to 20 ppm; 0 to 200%
  • Dissolved Oxygen Resolution0.1 ppm; 0.1% saturation
  • Dissolved Oxygen Accuracy±2%
  • Min Temperature (° C)5
  • Max Temperature (° C)45
  • Min Temperature (° F)41
  • Max Temperature (° F)113
  • InterfaceRS-232 and USB
  • WaterproofYes
  • Battery2 AA (included)
  • Battery life (hr)500
  • Width (cm)7.9375
  • Height (cm)6.35
  • Width (in)3 1/8
  • Height (in)2 1/2
  • DescriptionWaterproof pH/DO Portable Meter with Probes
  • Warranty3 year
3 Year WarrantyConformite EuropeenneIP 67

Includes

2 AA batteries.

Key Features

  • Optical DO sensor eliminates costly, tiresome membranes and solutions
  • Durable waterproof and dustproof design with IP67 rating
  • Barometric pressure and salinity correction
  • Memory for up to 500 data sets with USB or RS-232 output to PC
  • Built-in stand allows for easy benchtop use
  • 500 hours battery life, or optional universal power adapter
  • 3 year warranty

More About this Item

The PD 450 meter uses optical measurement technology offering fast response times and long-term stability with minimal maintenance for dissolved oxygen measurements. Plus the automatic temperature compensation (ATC), barometric pressure compensation, and salinity correction further ensure DO measurement accuracy. The large, backlit LCD lets you view pH, mV, % saturation, or mg/L (ppm) simultaneously with temperature.

Rugged, IP67-rated waterproof housing makes the meter ideal for plant or field testing. Additional features include ready (stability) indicator, smart averaging, hold function, calibration alarm, and battery-life indicator.

Perform data logging with manual or timed collection of up to 500 data sets with real-time clock time/date stamp. Order optional data cable for USB or RS-232 connectivity to a PC.

What's included: PD 450 meter, pH probe (35641-51), RDO probe (35640-51), sensor cap (35640-51), two AA batteries.

ACCESSORIES

Accessories

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat