Oakton PC2700 Benchtop Meter w/pH Electrode, Conductivity/ACT Probe, and Electrode Stand

Representative image only.
Oakton Item # RK-35414-00
Multiparameter benchtop meters are an all-in-one solution for laboratory research work
  • Advanced research-grade meter
  • Includes Ion concentration readings
  • 3 year warranty

$3,852.91CAD / Each

$242.37 - $349.19CAD / Each

Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypeBenchtop
  • Display typeLCD
  • ConnectionspH: BNC; Temperature: 2.5 mm
  • Min pH (pH)-2
  • Max pH (pH)20
  • pH Resolution0.001/0.01/0.1
  • pH Accuracy±0.002 pH
  • Buffer RecognitionDIN, NIST, and USA
  • CalibrationUp to 6 points
  • Min mV (mV)-2000
  • Max mV (mV)2000
  • mV Resolution0.1 or 1 mV
  • mV Accuracy±0.2 mV or 0.05%, whichever is greater
  • Min ISE0 to 19999 ppm
  • ISE Resolution0.001 ppm; 0.001 molar
  • ISE Accuracy±0.5% of reading
  • Max Conductivity0 - 500 mS
  • Conductivity Accuracy±1% FS
  • Min TDS0 to 500 ppt
  • Min Salinity0 to 80 ppt
  • Salinity Accuracy±1.0% FS
  • Resistivity Resolution0.01 mΩ; 0.01 or 0.1 Ω; 0.001 or 0.01 kΩ
  • Resistivity Accuracy±1% FS
  • Min Temperature (° C)-10
  • Max Temperature (° C)110
  • Min Temperature (° F)14
  • Max Temperature (° F)230
  • Temperature compensationAutomatic or manual
  • InterfaceRS-232
  • Power (VAC)110 to 240
  • Power (Hz)50 or 60
  • Width (cm)17.4625
  • Height (cm)6.985
  • Width (in)6 7/8
  • Length (in)6 1/8
  • Height (in)2 3/4
  • DescriptionMeter w/pH electrode, conductivity/temp probe, and electrode stand
  • Warranty3 year
3 Year WarrantyConformite EuropeenneISO9001:2000 Certified Supplier

Key Features

  • Advanced research-grade meter
  • Includes Ion concentration readings
  • 3 year warranty

More About this Item

Oakton PC 2700 meters feature pH auto-buffer recognition for both USA and NIST buffer sets, automatic or manual temperature compensation, and selectable temperature units. Previous calibration points, electrode slope and offset, and conductivity/TDS cell constants are stored in memory for quick and easy retrieval.

Meters offer an advanced user interface and advanced measurement capabilities. Meters meet GLP requirements—meters stamp all calibration and stored data with time/date. Nonvolatile memory stores up to 500 data sets to simplify documenting large volumes of pH, mV, ISE, conductivity, TDS, salinity, or resistivity measurements. Data is easily exported to a computer via RS-232 port. Order RS-232 and optional RS-232 to USB adapter separately.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat