Oakton Conductivity Calibration Pouches, 12.88 mS; 20/Pk
Representative image only.
Oakton Item # RK-35653-14
Sealed for freshness and easy travel
  • Tear off a corner, or the entire top—opens wide enough for all probe types
  • Notched corners for easy opening from either side
  • Calibrate to 12,888 µS or 12.888 mS
  • 1 year warranty
PRODUCT OPTIONS
$91.35CAD / Pkg of 20
SPECS & DESCRIPTION
Specifications & Description
  • Conductivity Value (µS)12880
  • Conductivity Value (mS)12.88
  • Volume (mL)20
  • Volume (oz)0.7
  • Volume20 pouches
  • Conductivity-to-TDS Calibration Values (ppm 442)11367
  • Conductivity-to-TDS Calibration Values (ppm KCl)7447
  • Conductivity-to-TDS Calibration Values (ppm NaCl)7230
  • DescriptionConductivity Calibration Pouches, 12.88 mS; Box of 20 Pouches
  • Warranty1 year
1 Year Warranty
Key Features
  • Tear off a corner, or the entire top—opens wide enough for all probe types
  • Notched corners for easy opening from either side
  • Calibrate to 12,888 µS or 12.888 mS
  • 1 year warranty
More About this Item
Single-use 20-mL pouches are sealed to ensure freshness—take these convenient standard solutions anywhere! Don't worry about transferring solution into another container—calibrate directly from the pouch.
A µS-to-ppm conversion table is printed on each pack for quick reference.