Oakton Conductivity and TDS Standard, 12,880 µS; 5 x 60 mL Bottles/Pk

Representative image only.
Oakton Item # RK-00653-37
Ideal for laboratory and field analysis
  • All Oakton Conductivity and TDS Standards are traceable to NIST standard reference materials
  • Solutions help you calibrate conductivity instruments to ensure accuracy
  • Standard values given at 25°C
  • Use when readings are expected to be around 12.88 mS/cm (12,880 µS/cm)
  • 1 year warranty

PRODUCT OPTIONS

$85.46CAD / Pkg of 5

SPECS & DESCRIPTION

Specifications & Description

  • Conductivity Value (µS)12880
  • Conductivity Value (mS)12.88
  • Volume (mL)60
  • Volume (oz)2
  • Conductivity-to-TDS Calibration Values (ppm 442)11367
  • Conductivity-to-TDS Calibration Values (ppm KCl)7447
  • Conductivity-to-TDS Calibration Values (ppm NaCl)7230
  • DescriptionConductivity and TDS Standard, 12,880 µS; 5 x 60 mL Bottles/Pk
  • Warranty1 year
1 Year Warranty

Key Features

  • All Oakton Conductivity and TDS Standards are traceable to NIST standard reference materials
  • Solutions help you calibrate conductivity instruments to ensure accuracy
  • Standard values given at 25°C
  • Use when readings are expected to be around 12.88 mS/cm (12,880 µS/cm)
  • 1 year warranty

More About this Item

KCl, NaCl and 442 conversions printed on the bottle for ease of use (excluded on 60-mL bottle, but included in insert). Temperature corrections are printed on the bottle, with certified values listed in tables and referenced to 25°C (excluded on 60-mL bottle, but included in insert). Manufactured under ISO 9000 quality guidelines and traceable to NIST standards. If unopened, shelf life is 1 year. If opened recommended shelf life is 4 months. Conductivity options range from 1 µS/cm to 200,000 µS/cm.

Convenient 60-mL bottles come in a pack of five—ideal for field use. Grab one when you're on the go!

Certificate of Analysis

Enter lot number
or click to view a Sample C of A

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat