Oakton Buffer Capsules, pH 7; 10/Pk
Representative image only.
Oakton Item # RK-35850-14
Fresh pH calibration solution anytime and anywhere
  • Each capsule mixes with 100-mL of deionized water to create a clear, 7.00 pH solution
  • 3 month warranty
PRODUCT OPTIONS
$37.13CAD / Pkg of 10
New Double Junction Waterproof Oakton pH Testr 50
SPECS & DESCRIPTION
Specifications & Description
  • Buffer Value7
  • Volume (mL)100
  • ColorClear
  • DescriptionBuffer Capsules, pH 7; 10/Pk
  • Warranty3 month
3 Month Warranty
Includes
Ten green-colored capsules.
Key Features
  • Each capsule mixes with 100-mL of deionized water to create a clear, 7.00 pH solution
  • 3 month warranty
More About this Item
Buffer salts inside capsules require 100 mL of deionized or distilled water to produce a clear pH calibration solution accurate to ±0.02 pH (at 25°C) with traceability to NIST standards. Capsules are easier to store and transport than prepared aqueous buffer solutions. Capsules are color-coded for easy identification if mixed.
Store buffer solution in an airtight container at room temperature for three weeks. Unused capsules have a five-year shelf life. Each vial contains 10 capsules.