This item is now discontinued. Please browse similar items
Traceable 8141 Static Suppressor
Representative image only.
Traceable Item # RK-98768-29
Reduce electrical noise from coiled wires or noisy environments
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithThin cables
  • DescriptionStatic Suppressor
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Static generated and radio frequency can affect any cable through the air or by physical contact. To protect against radio frequency, install a suppressor onto your thermometers cable to absorb radio frequency.