This item is now discontinued. Please browse similar items
North by Honeywell SSS Sleeves, 18"L, one pair
Representative image only.
North by Honeywell Mfr # SSS – Item # RK-86358-03
Resists a wide-range of chemicals
SPECS & DESCRIPTION
Specifications & Description
  • MaterialNorfoil®
  • SizeOne size fits all
  • Length (in)18
  • Length (cm)46
  • ColorGray
  • CuffElastic
  • SterileNo
  • DescriptionSleeves, 18\"L, one pair
More About this Item
This flexible lightweight sleeve is made of Norfoil, a laminate that resists permeation by a wide range of aromatic, chlorinated, aliphatic solvents, ketones, and esters. Ideal for lab or industrial use. Sleeve is one size fits all and features an elastic cuff at both ends.
Sleeves are one size fits all and feature an elastic cuff at both ends.