This item is now discontinued. Please browse similar items
North by Honeywell SSA Apron, 36" W x 44" L; 1/Pack
Representative image only.
North by Honeywell Mfr # SSA – Item # RK-86358-10
Superior resistance from chemical splash
SPECS & DESCRIPTION
Specifications & Description
  • Length (in)44
  • Length (cm)111.76
  • MaterialPolyethylene(PE)
  • Width (in)36
  • ColorSilver
  • Thickness (mil)2.7
  • DescriptionApron, 36\" W x 44\" L; 1/Pack
Conformite Europeenne
More About this Item
Lightweight apron is made of a five-layer polyethylene and ethylene vinyl alcohol copolymer construction that resists permeation of a wide range of ketones, esters, and aromatic, chlorinated, or aliphatic solvents. Apron measures 44"L x 36"W to maximize coverage. Adjust height and coverage using the attached ties. Aprons are recommended for use with corresponding Silver Shield sleeves and gloves. One size fits most.