This item is now discontinued. Please browse similar items
North by Honeywell 314LWR Powdered Latex finger cots, small, 1440/bag
Representative image only.
North by Honeywell Mfr # 314LWR/S – Item # RK-81605-32
Finger cots provide effective protection for delicate applications
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialLatex
  • Thickness (mil)3
  • SizeSmall
  • Length (in)2 3/4
  • Length (cm)7
  • ColorClear
  • ComplianceASTM D3772
  • SterileNo
  • DescriptionPowdered Latex finger cots, small, 1440/bag
  • Warranty1 year
1 Year Warranty
Key Features
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
  • 1 year warranty
More About this Item
Use when sensitivity and dexterity are needed. Bag of 1440.
Compliance: Meet ASTM D3772 for physical properties.