This item is now discontinued. Please browse similar items
Nor-Lake NSRF202WWW/0 Combination Refrigerator/Freezer, Solid, 10 cu ft; 115V/60Hz
Representative image only.
Nor-Lake Mfr # NSRF202WWW/0 – Item # RK-44174-26
Dual climate control in a space saving design
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionCombination Refrigerator/Freezer, Solid, 10 cu ft; 115V/60Hz
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Dual LED microprocessor display, one for the refrigerator and one for the freezer. Standard visual alarm on both the refrigerator and freezer alert you when the temperature gets too high or low. Bottom is composed of the freezer unit, while the top is the refrigerator. Door opening heater wires eliminate the formation of condensation on the doors.
Constructed of white, epoxy coated galvinized steel. Full length door pull handles and locks standard. Interior incandescent lighting for each section are activated by separate switches. Interior is white enamel, making for easy clean-up. Four casters are standard for easy movement.