This item is now discontinued. Please browse similar items
Non-Sterile Multichannel Disposable Solutions Basin
Representative image only.
Item # RK-24810-30
“V” shaped well design allows maximum reagent recovery
  • Easy to read graduation markings
  • Drip free pour-off spouts at every corner for spill free pouring
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • SterileNo
  • Max Volume (µL)55000
  • Min Volume (µL)0
  • Min Volume (mL)0
  • Max Volume (mL)55
  • Overall Length (in)5.7
  • Overall Length (mm)145
  • DescriptionNon-Sterile Multichannel Disposable Solutions Basin
  • Warranty1 year
1 Year Warranty
Key Features
  • Easy to read graduation markings
  • Drip free pour-off spouts at every corner for spill free pouring
  • 1 year warranty
More About this Item
Sterile reagent reservoirs are constructed of white polystyrene, providing easy to read graduation markings located on the top edge with a fill line on inner walls to indicate volume levels. The V-shaped bottom allows for efficient and nearly complete sample withdrawal.