Nichrome V Inoculating Loop with Insulated Handle, Calibrated, 26 Gauge, 1 µL
Representative image only.
Nichrome Mfr # 3730 – Item # RK-14203-25
Select, inoculate, or transfer microbiological samples
  • Nichrome wire composition cools quickly
  • Constructed of 26-gauge wire
  • 1 month warranty
PRODUCT OPTIONS
$43.32CAD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Loop Length (mm)44.5
  • Handle length (mm)152
  • Loop Length (in)1 3/4
  • Handle length (in)6
  • B and S Wire Gauge26
  • Contained Volume (µL)1
  • Handle TypeInsulated
  • ColorBlue
  • DescriptionV Inoculating Loop with Insulated Handle, Calibrated, 26 Gauge, 1 µL
  • Warranty1 month
1 Month Warranty
Key Features
  • Nichrome wire composition cools quickly
  • Constructed of 26-gauge wire
  • 1 month warranty
More About this Item
Ideal to streak various liquids, suspensions, bacteria, and other specimens on to culture media and/or petri dishes for purposes of study and growth. Each loop is calibrated to APHA standards for optimal accuracy and reproducibility. Choose from loops with or without handle. Order handle 14203-34 separately for loops without handle included.