This item is now discontinued. Please browse similar items
Newtex Industries ZFC High-Temp Finger Cots, 1000 F, One-Size; 12/Pk
Representative image only.
Newtex Industries Mfr # 2100004 – Item # RK-86364-02
  • Material resists up to 1000°F (538°C)
  • 1 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialCotton
  • SizeOne size fits all
  • ColorWhite
  • ComplianceEN 407
  • SterileNo
  • DescriptionHigh-Temp Finger Cots, 1000 F, One-Size; 12/Pk
  • Warranty1 month
1 Month Warranty
Key Features
  • Material resists up to 1000°F (538°C)
  • 1 month warranty
More About this Item
A safe alternative to asbestos, these gloves and finger cots are made of a durable cotton-backed, 11-ounce wool liner. Liner is sewn at the fingertips and double stitched with aramid thread throughout for strength and durability. One size fits all. Made in the USA. Compliance: European EN407 industrial standard
Note: The material resists up to 1000°F (538°C); operating temperature is 700°F (371°C) for longer exposure. These temperatures are dependent on duration, application, and environment. Actual use temperature may be lower.