This item is now discontinued. We recommend the following as suggested replacements: RK-53200-17
Neogen COLI9851BP Colitag™ Test Kit Blister Pack, P/A 100-mL Format; 100/Pk
Representative image only.
Mfr # COLI9851BP – Item # RK-53200-24
Quickly and confidently analyze drinking water
  • Colitag is a presence-absence and MPN enzyme substrate test
  • Detects as few as 1 MPN of total coliform or E. coli in a 100-mL water sample
  • US EPA approved; depended on in over 24 countries
  • Maximizes flexibility; use one test instead of three
SPECS & DESCRIPTION
Specifications & Description
  • TestColiform, E. coli
  • Tests Per Kit100
  • DescriptionColitag™ Test Kit Blister Pack, P/A 100-mL Format; 100/Pk
Key Features
  • Colitag is a presence-absence and MPN enzyme substrate test
  • Detects as few as 1 MPN of total coliform or E. coli in a 100-mL water sample
  • US EPA approved; depended on in over 24 countries
  • Maximizes flexibility; use one test instead of three
More About this Item
Enhance your laboratory efficiency—and ensure water purity—with Colitag, the revolutionary 16-48 hour water test. To use, simply add Colitag to a 100-mL sample, incubate, visually check for coliforms and illuminate with a UV lamp, as bright blue fluorescence indicates a positive E. coli sample.