This item is now discontinued. Please browse similar items
Motic Instruments Microscope Camera Adaptor, C-Mount, 1X, No Lens
Representative image only.
Motic Instruments Mfr # 1101001901771 – Item # RK-48412-01
Easily connect your trinocular microscope to a camera
  • Compatible with Motic® trinocular microscopes
  • Compatible with Motic BA Series, AE31E/AE2000, and SMZ-161/171 trinocular microscopes
Include Camera Today...
Motic® Moticam Microscope Cameras
$640.56 - $2,187.98CAD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithMotic BA Series, AE31E/AE2000, and SMZ-161/171 trinocular microscopes
  • ConnectionsC-Mount
  • DescriptionMicroscope Camera Adaptor, C-Mount, 1X, No Lens
Key Features
  • Compatible with Motic® trinocular microscopes
  • Compatible with Motic BA Series, AE31E/AE2000, and SMZ-161/171 trinocular microscopes
More About this Item
In order to use your microscope camera with the trinocular port on your microscope, you need a C-mount adapter to join the microscope and camera together. The C-mount adapter that is needed will depend on your microscope, as well as your camera.