This item is now discontinued. We recommend the following as suggested replacements: RK-07571-02
Masterflex E/S Portable Sampler, 115 VAC
Representative image only.
Masterflex Item # RK-07571-00
Economical choice for basic sampling applications
  • Features a single-turn potentiometer for speed control, ON indicator, and three-position power/reversing switch
  • Reversible flow and prime/purge function
  • Uses Masterflex® L/S® standard, Quick Load®, PTFE tubing, or Easy-Load® pump heads (order separately)
  • Floats up to 30 minutes if dropped in water when case is closed and latched!
  • IP54-rated control panel protects against
SPECS & DESCRIPTION
Specifications & Description
  • Sampling PumpPeristaltic
  • Max Lift (ft)26
  • Max Lift (m)7.9
  • Power115 VAC, 50/60 Hz
  • DescriptionE/S portable sampler, 115 VAC
Key Features
  • Features a single-turn potentiometer for speed control, ON indicator, and three-position power/reversing switch
  • Reversible flow and prime/purge function
  • Uses Masterflex® L/S® standard, Quick Load®, PTFE tubing, or Easy-Load® pump heads (order separately)
  • Floats up to 30 minutes if dropped in water when case is closed and latched!
  • IP54-rated control panel protects against
  • inclement weather and splashing water
  • Rechargeable battery — ideal for field applications
More About this Item
The Masterflex® E/S™ portable sampler, part of our environmental sampling series from Masterflex®, is designed for the ultimate in portability and versatility. Run up to four hours on the self-contained 12 VDC rechargeable battery, or indefinitely from any AC or 12 VDC power supply. Charge the internal battery from an AC power supply or a DC voltage source of lower voltaic potential than the battery itself. The variable-speed drive has a top speed of 400 rpm, allowing you to adjust the flow rate to the needs of your application.
Pump is encased in a high-visibility yellow housing for easy recognition and recovery in the field. Unit measures 16"H x 11-1/3"W x 10-1/2"D. What's included: instruction manual and AC adapter with 12-ft cable. Order automotive power adapter separately to run off of a DC power supply.
The Masterflex® L/S® standard, Quick Load® or Easy-Load® pump head must be ordered separately - it is NOT included.