Micropump 81094 Drive for A-Mount pump heads, explosion-proof (XPRF), 3150 rpm, 115/230 VAC

Representative image only.
Micropump Mfr # 85734 – Item # RK-07144-95
A variety of low-cost drives for numerous applications
  • Magnetic coupling minimizes friction—all horsepower is transmitted directly to the gears
  • Choose either an explosion-proof or air-powered model for safe pumping in hazardous areas
  • Hundreds of hours of dependable leakproof service

PRODUCT OPTIONS

$2,370.68CAD / Each

$212.17CAD / Each

for an additional
$212.17CAD / Each
Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • Max Motor RPM (rpm)3150
  • Motor Enclosure TypePSC
  • Power (VAC)115 / 230
  • Power (amps)1.8
  • Duty Cycle ContinuousYes
  • Max Temperature (° F)140
  • Min Temperature (° F)40
  • Max Temperature (° C)60
  • Min Temperature (° C)4
  • Width (in)6
  • Width (cm)15.2
  • Length (in)11
  • Length (cm)27.9
  • Height (in)5.25
  • Height (cm)13.3
  • Motor hp0.17
  • DescriptionDrive for A-Mount pump heads, explosion-proof (XPRF), 3150 rpm, 115/230 VAC

Key Features

  • Magnetic coupling minimizes friction—all horsepower is transmitted directly to the gears
  • Choose either an explosion-proof or air-powered model for safe pumping in hazardous areas
  • Hundreds of hours of dependable leakproof service

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat