This item is now discontinued. Please browse similar items
Ansell Microflex® Nitrile Cleanroom Glove, ISO Class 4 compliant, medium, 100/bag
Representative image only.
Ansell Mfr # CE4-313-M – Item # RK-86313-46
Low surface particulates, ionic extractables, and non-volatile residues
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
  • 1 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialNitrile
  • Thickness (mil)5.2
  • SizeMedium
  • Length (in)12
  • Length (cm)30.5
  • ColorClear
  • ComplianceISO Class 4
  • SterileNo
  • DescriptionNitrile Cleanroom Glove, ISO Class 4 compliant, medium, 100/bag
  • Warranty1 month
1 Month WarrantyConformite Europeenne
Key Features
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
  • 1 month warranty
More About this Item
Cleanroom gloves are washed in a microprocessor-controlled washer/dryer system to insure low residues. Micro-textured surface provides wet or dry grip. Disposable; for single use only. Ambidextrous.
Compliance: ISO Class 4 or 5 (see table).
Nitrile material alleviates adverse reactions to latex protein and also features a surface resistivity of 1011 for static decay.