This item is now discontinued. Please browse similar items
Mettler Toledo Flow pH Electrode; S7
Representative image only.
Mettler Toledo Mfr # 59902917 – Item # RK-35902-06
Expertly-engineered probes to cover a wide range of applications
  • Cable not included
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsMoving samples
  • Electrode Body MaterialGlass
  • Reference JunctionDouble ceramic
  • Reference ElectrolyteNon-refillable gel
  • Sensing Glass TypeU - Universal for standard applications
  • Min pH (pH)0
  • Max pH (pH)11
  • Min Temperature (° C)0
  • Max Temperature (° C)80
  • ATC Element
  • ConnectorS7
  • Diameter (mm)7
  • Length (mm)40
  • DescriptionpH Electrode; S7
Key Features
  • Cable not included
More About this Item
These ISM® - Intelligent Sensor Management electrodes models store calibration data and automatically transfer the data to your meter. Choose open junction versions for dirty and complex samples. Cables not included with all electrodes. For additional versatility—select the one that connects to your meter.
Select the appropriate sensor cable for your application and meter—BNC, waterproof BNC, DIN, Lemo OO, and wire-only cables available.