Mettler Toledo EasyPlus Easy pH Acid/Base Titrator, 20 mL; 100-240 VAC, 50/60 Hz

Representative image only.
Mettler Toledo Mfr # 30060041 – Item # RK-94190-02
Complete packages designed to simplify key applications
  • Simple apps-based user interface for easy operation
  • Precise burettes and sensors ensure system accuracy
  • Data easily transferred to PC with optional EasyDirect software
  • Ideal for acidity of beverages, alkalinity of cleaning reagents, free fatty acid content of cooking oils, and alkalinity and acidity of water
  • Performs direct, back compensated, and blank compensated titrations
  • Offers choice of aqueous and non-aqueous pH electrodes
  • 1 year warranty

PRODUCT OPTIONS

Call For Price
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • UnitspH, mV
  • Display TypeTouchscreen
  • Display Resolution4870 x 272 pixels
  • Min pH (pH)0
  • Max pH (pH)14
  • pH Repeatability± 0.5% RDS
  • pH Resolution0.1
  • Max mV (mV)2000
  • mV Resolution0.1
  • Min Temperature (° C)-5;0
  • Max Temperature (DegreeC)100;105
  • Capacity (mL)20
  • OutputUSB
  • Data LoggingYes
  • Power (VAC)100 to 240
  • Power (Hz)50/60
  • Length (in)8 21/32;6 11/16
  • Width (in)8 21/32;6 11/16
  • Height (in)13 3/4
  • Length (cm)22;17
  • Width (cm)22;17
  • Height (cm)35
  • DescriptionAcid/Base Titrator, 20 mL; 100-240 VAC, 50/60 Hz
  • Warranty1 Year
1 Year WarrantyISO9001 Certified Supplier

Includes

Aqueous pH sensor, 20 mL burette, tubing set, stirrer, titration head, drying tube, valve, bottle head, insert set for head, and burette cover.

Key Features

  • Simple apps-based user interface for easy operation
  • Precise burettes and sensors ensure system accuracy
  • Data easily transferred to PC with optional EasyDirect software
  • Ideal for acidity of beverages, alkalinity of cleaning reagents, free fatty acid content of cooking oils, and alkalinity and acidity of water
  • Performs direct, back compensated, and blank compensated titrations
  • Offers choice of aqueous and non-aqueous pH electrodes
  • 1 year warranty

More About this Item

Affordable automatic titration packages are designed with everything you need for titrations--the ideal replacement for laboratories still performing manual titrations! Although small in size, these quality titrators provide highly accurate and consistent results.

Operation is simple and intuitive thanks to the smartphone-based Apps-driven user interface; use it to control all major functions. Easily perform basic titrations and receive automatic results from predefined formulas. Setup and tutorial with navigation is available in 14 languages. Diagnostic tools assist with the most common issues to minimize downtime.

The pH titrator is designed to determine acidity in juices, milk, and wine, p-and m-values in water, and more.The iTitrate™ built-in user interface requires only a few parameters to be set before the instrument is ready for acid/base titration.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat