This item is now discontinued. Please browse similar items
Mettler Toledo 30460841 Left Draft Shield Door for XPR/XSR Analytical Balances
Representative image only.
Mettler Toledo Mfr # 30460841 – Item # RK-11337-82
Optimize performance of your Mettler Toledo touch XPR and XSR Excellence precision balances
  • Keep your balance running smoothly with these parts and accessories
  • Protect weighing samples from surrounding influences by minimizing air turbulence
  • Ensures faster stabilization
  • Slotted shield allows easy placement of automatic feeder LV12
  • 3 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Accessory TypeLeft Draft Shield Door
  • DescriptionLeft Draft Shield Door for XPR/XSR Analytical Balances
  • Warranty3 month
3 Month Warranty
Key Features
  • Keep your balance running smoothly with these parts and accessories
  • Protect weighing samples from surrounding influences by minimizing air turbulence
  • Ensures faster stabilization
  • Slotted shield allows easy placement of automatic feeder LV12
  • 3 month warranty
More About this Item
Choose from a wide range of accessories suitable for your balance including covers, draft shields, ionizers, and more.