This item is now discontinued. Please browse similar items
Met One Instruments, Inc. GT-321 Particle Monitor, Portable
Representative image only.
Met One Instruments, Inc. Mfr # GT-321 – Item # RK-18460-00
Monitor particulate concentrations in your clean room
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Particle Size (micron)0.3, 0.5, 1.0, 2.0, 5.0
  • Max Temperature (° F)122
  • Min Temperature (° F)32
  • Max Temperature (° C)50
  • Min Temperature (° C)0
  • Max Flow Rate (CFM)0.1
  • Max Flow Rate (LPM)2.83
  • DisplayLCD
  • Light SourceLaser diode
  • OutputRS-232
  • PowerRechargeable NiMH battery with 100 to 240 VAC charger
  • Battery Life8 hours intermittent use; 5 hours continuous use
  • DescriptionInstruments, Inc. Particle Monitor, Portable
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Key Features
  • 1 year warranty
More About this Item
Handheld particle monitor features five size ranges that can be selected from the front panel. Monitor detects 0.3 ìm or greater sized particles at concentrations from 0 to 3,000,000 particles per cubic foot. The built-in rate counter provides a series of six-second count periods, each period is extrapolated to provide a concentration per sample volume. Results are shown on the 16-character LCD.