This item is now discontinued. Please browse similar items
Meiji Techno Stereozoom microscope system with photo tube, 7x to 45x continuous magnification
Representative image only.
Meiji Techno Mfr # EMZ-5TR + PBH+MA502 – Item # RK-48402-00
Convenient, preassembled microscope systems provide superb quality and easy ordering
  • Model 48402-00 includes:
  • Stereozoom body 48402-25
  • Standard stand 48404-20
  • 10x eyepieces 48402-58
  • Limited Lifetime Warranty
SPECS & DESCRIPTION
Specifications & Description
  • Magnification7x to 45x
  • Maximum Magnification45x
  • Head ConfigurationTrinocular
  • Objectives0.7x to 4.5x
  • Eyepiece Diameter (mm)34
  • Eyepiece Magnification10x
  • EyepiecesWidefield
  • Illuminator(s)Top and bottom
  • Stand TypeStandard
  • Power (VAC)115
  • DescriptionStereozoom microscope system with photo tube, 7x to 45x continuous magnification
  • WarrantyLTD Life Time
Conformite EuropeenneLimited Lifetime Warranty
Key Features
  • Model 48402-00 includes:
  • Stereozoom body 48402-25
  • Standard stand 48404-20
  • 10x eyepieces 48402-58
  • Limited Lifetime Warranty
More About this Item
For your added convenience, we also offer preassembled microscope systems that include a microscope body, stand, and eyepieces. All eyepieces have diameters of 34 mm OD x 25 mm ID.