This item is now discontinued. Please browse similar items
Meiji Techno MX7000 Metallurgical Binocular Microscope; Brightfield/Incident
Representative image only.
Meiji Techno Mfr # MT7000 – Item # RK-48405-21
View metals, plastics, ceramics, paint, glass, crystals, and semiconductor components
  • Plan Epi optics provide clear images, excellent color rendering, and superior flatness of field
  • Limited Lifetime Warranty
SPECS & DESCRIPTION
Specifications & Description
  • Magnification5x/10x/20x/50x
  • Maximum Magnification50x
  • Head ConfigurationBinocular
  • ObjectivesEpi-plan, infinity-corrected
  • Eyepiece Magnification10x
  • EyepiecesPaired DIN
  • Stage Dimensions191 x 126 mm
  • Illuminator(s)Brightfield/Incident
  • Power (VAC)110 / 240
  • DescriptionMetallurgical Binocular Microscope; Brightfield/Incident
  • WarrantyLTD Life Time
Limited Lifetime Warranty
Key Features
  • Plan Epi optics provide clear images, excellent color rendering, and superior flatness of field
  • Limited Lifetime Warranty
More About this Item
Metallurgical microscopes illuminate the specimen through the objective lens — ideal for specimens that don't transmit light, such as metals. Comfortable Siedentopf viewing heads have 30'' inclined eyetubes with one adjustable diopter and one fixed diopter. The interpupillary distance adjustable from 53 to 75 mm. Ergonomic coaxial focus controls have tension adjustment and a safety autostop. Koehler illumination allows for effective contrast control. The illuminator features field iris, aperature iris, and filter slots. Filter sets are included.
Microscopes with brightfield/incident illumination are ideal for everyday use. Features Epi-Plan infinity corrected optics and scratch-resistant ceramic coated stage. Each model includes polarized light observation mode.