This item is now discontinued. Please browse similar items
Meiji Techno EMZ-5TR Stereozoom microscope bodies; objectives, 0.7x to 4.5x; photo tube included
Representative image only.
Meiji Techno Mfr # EMZ-5TR – Item # RK-48402-25
  • Limited Lifetime Warranty
SPECS & DESCRIPTION
Specifications & Description
  • Magnification0.7 to 4.5x
  • Maximum Magnification4.5x
  • Head ConfigurationTrinocular
  • Objectives0.7x to 4.5x
  • DescriptionStereozoom microscope bodies; objectives, 0.7x to 4.5x; photo tube included
  • WarrantyLTD Life Time
Limited Lifetime Warranty
Key Features
  • Limited Lifetime Warranty
More About this Item
Get crisp, high-resolution images with excellent stereoscopic viewing. The 45° inclined head has adjustable diopter and interpupillary controls for easy observing. Rotate body a full 360° and lock into any position. The eyepiece diameters on all models are 25 mm ID x 34 mm OD.
Model 48402-25 has convenient side-mounted zoom controls for increased efficiency. Also features a vertically mounted photo tube for use with a video camera that has a C-mount adapter—ideal for documenting, teaching, or recording images.