This item is now discontinued. Please browse similar items
Megger ICLAMP Current Measuring Clamp
Representative image only.
Megger Mfr # 1001-012 – Item # RK-20004-61
  • Permits separation of test currents in a parallel-grounded system
  • Enables grounds to be tested without de-energizing the system
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionCurrent Measuring Clamp
  • Warranty1 Year
1 Year Warranty
Key Features
  • Permits separation of test currents in a parallel-grounded system
  • Enables grounds to be tested without de-energizing the system
  • 1 year warranty
More About this Item
Choose from these quality parts and accessories to meet your application needs.
Use the current clamp along with the Megger ground resistance tester to perform rod technique ground resistance tests.