This item is now discontinued. View the direct replacement: RK-08491-02 or browse similar items
Measurement Specialties 401 400 Series Thermistor General Purpose Probe
Representative image only.
Measurement Specialties Mfr # 401 – Item # RK-08430-00
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Probe TypeThermistor
  • Min Temperature (° F)-40
  • Max Temperature (° F)212
  • Min Temperature (° C)-40
  • Max Temperature (° C)100
  • Time Constant7 seconds
  • Connector TypePhono plug
  • Sheath Length (in)0.3125
  • Sheath Diameter (in)0.1406
  • Description400 Series Thermistor General Purpose Probe
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Probes are accurate to +/- 0.1 deg C from 0 to 70 deg C. Electrically isolated probes come with a nondetachable 10-ft lead with phone plug (except as noted).
Water-resistant junctions between probe and lead should not be immersed; detachable leads are not water resistant. Note: Time constant is estimate time required to reach 63.2% of a new reading. To determine how long it will take to reach 99% of a new reading multiply the time constant by five.
1/4" or 6.350 mm phono plug connection
As of 11/2016 the vendor no longer has CE approval on this product. The product has not changed.