Masterflex Y-connector, polypropylene, L/S 15-25, 10/pack SKU 3061451

Representative image only.
Masterflex Item # RK-30614-51
  • Designed to tight tolerances
  • Made from virgin materials

PRODUCT OPTIONS

$43.02CAD / Pkg of 10

SPECS & DESCRIPTION

Specifications & Description

  • Fitting MaterialPolypropylene
  • Sterilization MethodAutoclave or ethylene oxide (ETO)
  • Connection TypeBarb
  • Barb Size / Tubing Id (Port 1) (in)3/16
  • Barb Size / Tubing Id (Port 2) (in)3/16
  • Barb Size / Tubing Id (Port 3) (in)3/16
  • DescriptionY-connector, polypropylene, L/S 15-25, 10/pack
Certificate of Compliance

Key Features

  • Designed to tight tolerances
  • Made from virgin materials

More About this Item

Ensure a tight seal every time with Masterflex fittings. Designed to tight tolerances, these fittings match perfectly to your tubing to ensure smooth flow and a proper fit. Fittings are available in four materials and are available in packs of 10.
High-Density Polyethylene (HDPE): Very good chemical resistance. Temp range: -65 to 150°F (-53 to 65°C). Sterilize by ethylene oxide only.
Nylon: Good chemical resistance. Temp range: -50 to 250°F (-46 to 121°C). Sterilize by ethylene oxide only.
Polypropylene: Very good chemical resistance. Temp range: 32 to 275°F (0 to 135°C). Sterilize by ethylene oxide or autoclave.
Kynar®: Excellent chemical resistance. Temp range: -80 to 265°F (-62 to 129°

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat

Alternatives

Alternatives