Masterflex Variable-Area Flowmeter with Valve, Direct Reading, Brass Fittings, 150-mm Scale; 200 mL/min Air
Representative image only.
Masterflex Item # RK-32004-06
No more eye strain - front shield magnifies scale 16 percent for easier readings
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
  • Scale will not wear off—made of fused ceramic for durability
  • Ensure accurate readings - vertical tangential locator with hairline accuracies
  • Aluminum fittings are economical and good for general use with noncorrosive gases and liquid
  • 316 Stainless Steel (SS) fittings withstand higher temperatures and pressures, and features excellent chemical compatibility
PRODUCT OPTIONS
$331.07CAD / Each
BUY MORE & SAVE
  • 1-3$331.07CAD / Each
  • 4-9$297.96CAD / Each
  • 10-19$281.41CAD / Each
  • 20+$264.86CAD / Each
$400.76CAD / Each
for an additional
$400.76CAD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy±5% full-scale
  • Repeatability±0.25% full-scale
  • Max Flow Rate (mL/min)200
  • Material Flow TubeBorosilicate glass
  • Material FittingChrome-plated brass
  • Material ValveBrass
  • Material O-RingBuna-N
  • Material Float316 SS
  • Material HousingBrass
  • ValveYes
  • Max Operating Temperature (° F)250
  • Max Operating Temperature (° C)121
  • Process Connection (NPT(F))0
  • Fluid TypeAir
  • Description150-mm Direct Reading Flowmeter, Br/SS Float; Air, 200 mL/min, Valved
Includes
150-mesh inlet screen.
Key Features
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
  • Scale will not wear off—made of fused ceramic for durability
  • Ensure accurate readings - vertical tangential locator with hairline accuracies
  • Aluminum fittings are economical and good for general use with noncorrosive gases and liquid
  • 316 Stainless Steel (SS) fittings withstand higher temperatures and pressures, and features excellent chemical compatibility
  • Chrome-plated brass is durable and easy to clean
  • SS float has a shiny silver finish and a density of 8.04 g/mL
  • Max flow for air is 200 mL/min
More About this Item
This full range of direct-reading flowmeters provide a robust solution for your laboratory that eliminates the need for reading conversions. Valved flowmeters have a valve mounted at the inlet (bottom) of the flowmeter. This arrangement is typically used for positive pressure applications for liquids and gases. The valve can be reconfigured to mount at the outlet (top), often for vacuum applications.