Masterflex Variable-Area Flowmeter Kit with Valve, Direct Reading, Acrylic Housing and Brass Fittings; 16 L/min Air
Representative image only.
Masterflex Item # RK-34500-46
Seven interchangeable scales for routine gases and water offer flexibility
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
  • Designed with stable, easy-to-read float for quick measurement readings
  • Interchangeable scales offer English and metric scales
  • Use with multiple routine gases for laboratory and industrial applications
  • Easy disassembly and assembly for cleaning
PRODUCT OPTIONS
$119.64CAD / Each
BUY MORE & SAVE
  • 1-3$119.64CAD / Each
  • 4-9$107.68CAD / Each
  • 10-19$101.69CAD / Each
  • 20+$95.71CAD / Each
$400.76CAD / Each
for an additional
$400.76CAD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy±5% full-scale
  • Max Pressure (PSI)100
  • Max Air Flow Rate (mL/min)16000
  • Max Flow Rate (mL/min)16000
  • Material FittingBrass
  • Material O-RingViton®
  • Material FloatSS
  • Material Housingacrylic
  • Valvebrass
  • Max Operating Temperature (° F)150
  • Max Operating Temperature (° C)65
  • Process Connection (NPT(F))0
  • DescriptionFlowmeter Kit, Brass Fittings; 16 LPM Air With Valve
Key Features
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
  • Designed with stable, easy-to-read float for quick measurement readings
  • Interchangeable scales offer English and metric scales
  • Use with multiple routine gases for laboratory and industrial applications
  • Easy disassembly and assembly for cleaning
More About this Item
These easy-to-read acrylic flowmeter kits come with seven interchangeable back plates for routine gases air, argon, nitrogen, helium, carbon dioxide, oxygen, and water. Meters are offered with or without built-in low hysteresis needle valves. Meters without valves can be panel mounted or used in partial or full in-line mounted configurations.