Masterflex Variable-Area Flowmeter, Direct Reading, Acrylic Housing and Brass Fittings with valve, 16 L/min Air
Representative image only.
Masterflex Item # RK-68560-18
Designed for labs and plant processes where multiple gases are used
  • Easy to read English or metric scales
  • Interchangeable scales for routine gases
  • Customer convertible panel to partial or full in-line mounted configurations
  • Panel mounting by means of hex nuts
  • Easy disassembly and assembly for cleaning
PRODUCT OPTIONS
$90.10CAD / Each
BUY MORE & SAVE
  • 1-3$90.10CAD / Each
  • 4-9$81.09CAD / Each
  • 10-19$76.59CAD / Each
  • 20+$72.08CAD / Each
$95.77 - $400.76CAD / Each
for an additional
$95.77CAD / Each
for an additional
$400.76CAD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy±5% full-scale
  • Max Pressure (PSI)100
  • Max Flow Rate (LPM)16
  • Material FittingBrass
  • Material O-RingViton®
  • Material HousingAcrylic
  • ValveYes
  • Wetted MaterialsBrass
  • Max Operating Temperature (° F)150
  • Max Operating Temperature (° C)65
  • Media TypeAir
  • DescriptionAcrylic Flowmeter w/ Valve for Air, Brass, 16 LPM
Key Features
  • Easy to read English or metric scales
  • Interchangeable scales for routine gases
  • Customer convertible panel to partial or full in-line mounted configurations
  • Panel mounting by means of hex nuts
  • Easy disassembly and assembly for cleaning
  • Flowmeter features brass fittings and Viton® o-ring
More About this Item
Versatile acrylic flowmeters are designed specifically for air. Flowmeters are offered with or without built-in low hysteresis needle valves. Panel mount flowmeters without valves or use in partial or full in-line mounted configurations. Flowmeters feature a stable, easy-to-read float for quick measurement readings.