Masterflex Single-Use Sterile Sampling Bottle Assembly, PETG Bottle; 2 L

Representative image only.
Masterflex Item # RK-95644-08
Safely transfer, sample, and store biopharmaceutical products and reagents
  • Secure, single-piece cap/tubing assembly eliminates contamination and product loss due to lose connections
  • Assemblies are pre-sterilized by gamma irradiation—eliminates labor and validation costs associated with assembly and cleaning
  • Designed with 2 L bottle volume

PRODUCT OPTIONS

$287.31CAD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (mL)2000
  • Capacity (L)2
  • Capacity (oz)64
  • MaterialPolyethylene Terephthalate (PET)
  • Cap / Stopper Size38 mm
  • Cap ColorNatural
  • Bottle ColorTransparent
  • DescriptionSterilized Sampling Bottle Assembly, 2 L

Includes

clear PETG bottle, 38-mm cap, C-Flex® tube/liner assembly with 1/8" ID x 1/4" OD tubing, and 0.2-micron vent filter.

Key Features

  • Secure, single-piece cap/tubing assembly eliminates contamination and product loss due to lose connections
  • Assemblies are pre-sterilized by gamma irradiation—eliminates labor and validation costs associated with assembly and cleaning
  • Designed with 2 L bottle volume

More About this Item

Cap assembly includes a seamless liner with premolded C-Flex® fill and vent tubes to create an unobstructed fluid path. The 18" fill tube is plugged at the terminal end to maintain assembly sterility until use. Vent tube includes a 0.2-micron PTFE vent filter to prevent infiltration by contaminants. Vent filter is hydrophobic to guard against loss of costly product or media during filling or emptying. The one-piece liner eliminates the risk of bacterial entrapment and contamination and forms an elastomeric seal between cap and bottle rim.

C-Flex tubing is manufactured according to cGMP and meets USP Class VI and USDA requirements. Tubing can withstand temperatures from –100 to 275°F (–73 to 135°C). C-Flex tubing has exceptional tensile and tear strength, extremely low extractables, and excellent biocompatibility. The smooth inner surface reduces absorption and protein binding.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat