This item is now discontinued. Please browse similar items
Fitting, Black Polypropylene, Straight, Male Luer Lock to Threaded Adapter, 1/4" ID x 1/4" NPT(M); 10/PK
Representative image only.
Mfr # MAKE UP – Item # RK-30505-52
Easily transition between luer fittings and UNF or NPT threads
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Connection Type: 1/4" ID x 1/4" NPT(M)
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene
  • Number of Ports2
  • Connection Type-Port 1Male luer lock
  • Connection Type-Port 20.25-18 NPT(M)
  • DescriptionMale luer lock x 1/4\" NPT, Black Polypropylene, 10 Pack
Key Features
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Connection Type: 1/4" ID x 1/4" NPT(M)
More About this Item
These adapters transition between luer fittings and UNF or NPT threads. Both male and female luer connections are available. All threaded connections are male thread.