This item is now discontinued. Please browse similar items
Marsh Bellofram 960-333-000 High-Flow Regulator, 0 to 60 psi, 200 scfm
Representative image only.
Marsh Bellofram Mfr # 960-333-000 – Item # RK-68825-96
Designed for applications that require high flow and accurate pressure control
  • Rolling diaphragm provides low-friction, long life service
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Min Exhaust Flow Rate (SCFM)14
  • Forward Flow Capacity (SCFM)Up to 200
  • Forward Flow Capacity (LPM)Up to 5660
  • Min Operating Temperature (° F)-20
  • Max Operating Temperature (° F)180
  • Min Operating Temperature (° C)-29
  • Max Operating Temperature (° C)82
  • Supply Pressure Effect< 0.35 psi per 100 psi
  • Max Supply Pressure (PSI)400
  • Sensitivity (in H₂O)1
  • Bleed Rate (SCFH)12
  • DescriptionHigh-Flow Regulator, 0 to 60 psi, 200 scfm
  • Warranty1 year
1 Year Warranty
Key Features
  • Rolling diaphragm provides low-friction, long life service
  • 1 year warranty
More About this Item
With a maximum supply pressure of up to 400 psi (27.6 bar) and output ranges up to 125 psi (8.6 bar), these pressure regulators can achieve flow rates of well over 200 SCFM. Once set to a desired output pressure, these direct-acting diaphragm-operated regulators maintain the setting permanently. The rugged die-cast zinc and aluminum housings are pressure tested to assure safe operation, and are designed to withstand harsh and abusive environments. Can be panel or pipe mounted.