Mark-10 MR51-50Z Torque Sensor, 50 ozFin / 3600 gFcm / 36 kgFmm / 350 Nmm / 35 Ncm
Representative image only.
Mark-10 Mfr # MR51-50Z – Item # RK-25302-32
Outstanding overload protection
  • Interchangeable chucks (order separately)
  • Warranty: 3 years
  • Outstanding overload protection
  • Interchangeable chucks (order separately)
  • Warranty: 3 years
PRODUCT OPTIONS
$1,407.09CAD / Each
$406.66 - $514.21CAD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (oz-in)50
  • Capacity (g-cm)3600
  • Resolution (oz-in)0.02
  • Resolution (g-cm)2
  • Resolution (N-cm)0.02
  • Accuracy±0.20% of full scale + indicator
  • DescriptionPlug & Test Universal Torque Sensor, Interchange Grips; 35 N cm
3 Year WarrantyConformite Europeenne
Key Features
  • Interchangeable chucks (order separately)
  • Warranty: 3 years
  • Outstanding overload protection
  • Interchangeable chucks (order separately)
  • Warranty: 3 years
More About this Item
A great solution for general bi-directional torque testing applications requiring different sample sizes. Three interchangeable chuck capacities and bit holder are available separately.
A great solution for general bi-directional torque testing applications requiring different sample sizes. Three interchangeable chuck capacities and bit holder are available separately.